Learn More
Sharpsguard™ Eco 0.5 Waste Container
GREENER_CHOICE
Product Code. 17734844
Change view
Click to view available options
Capacity (Metric):
0.4 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
17734844 0.4 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17734844 Supplier Sharpsguard™ Supplier No. DD442YLR
Only null left
Add to basket
Add to basket
The new Sharpsguard™ eco yellow web containers, made using at least 40% recycled plastic that is used for the disposal of sharps that have been contaminated with medicines, but not cytotoxic
  • Sharpsguard™ eco yellow 0.5 with web opening, is ideal for community healthcare workers and easily fitted into a bag or glove compartment.
  • This container is supplied with lid assembled.
  • Apart from a difference in shade of color and an added 'R' on the end of the product code, there is no difference to the safe and easy to use range that you know and trust.
  • This container is for disposal by incineration or other authorized process and is designed for the consignment of waste categorized by EWC codes: 18-01-01, 18-01-03, 18-01-09, 18-02-01, 18-02-02, 18-02-08.
Specifications
Capacity (Metric) 0.4 L
Color Yellow
Dimensions (L x W x H) 79 x 45 x 206 mm
Opening Size (Metric) 26 x 30 mm
Quantity 1 Ea.
Weight (Metric) 88 g