Learn More
Bellingham Stanley™ Eclipse Professional Grade Optical Hand-Held Water-in-Honey Refractometer
Product Code. 12918544
Click to view available options
Unit Size:
Each
Product Code. 12918544
Brand: Bellingham Stanley™ 4527
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For ‘on the spot’ determination of Refractive Index (RI) of liquids or the concentration of solutions
Hand held refractometers manufactured to the highest specification using only the best materials and optical components. Sealed against the ingress of water, they can be used in the harshest of environments including cold storage facilities and wet production halls. The large body housing and rubber hand grip make the instrument easy to handle. A number of special scales are available for general use and special models are available, such as the “low volume” versions which facilitate measurement of samples of less than 1μL, used by botanists and zoologists for measuring nectar.
  • Ideal for factory or general applications
  • Extremely high build quality
  • All metal “sealed” Construction to IP65
  • Rubber handgrip for easy handling
TRUSTED_SUSTAINABILITY
Specifications
Type Eclipse Professional Grade Optical Hand Held Refractometer
Scale Range 10 to 30% Water-in-Honey
For Use With (Application) ‘On the spot’ determination of Refractive Index (RI) of liquids or the concentration of solutions
IP Rating IP65