Learn More
Bellingham + Stanley™ Eclipse Hand Held Refractometer
Product Code. 11924229
Change view
Click to view available options
Increments:
0.1
0.2
0.5
Scale Type:
Brix
Low Volume Brix
Refractive Index
Unit Size:
Each
Scale Range:
0 to 15 Brix
0 to 30 Brix
0 to 50 Brix
28 to 65 Brix
45 to 80 Brix
72 to 95 Brix
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Increments Scale Type Unit Size Scale Range
11924229 0.2 Refractive Index Each -
12303519 0.1 Brix Each 0 to 15 Brix
12313519 0.2 Brix Each 0 to 30 Brix
12353519 0.2 Brix Each 45 to 80 Brix
12343519 0.2 Brix Each 28 to 65 Brix
12393529 0.5 Low Volume Brix Each 0 to 50 Brix
12303539 0.2 Low Volume Brix Each 45 to 80 Brix
12333519 0.5 Brix Each 0 to 50 Brix
12363519 0.2 Brix Each 72 to 95 Brix
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Product Code. 11924229 Supplier Bellingham + Stanley™ Supplier No. 4541
Estimated Shipment: 15-12-2026
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Manufactured to the highest specification using only the best materials and optical components. Bellingham + Stanley™ Eclipse Hand Held Refractometer is a low-cost, portable instrument used for on-the-spot determination of Refractive Index of liquids or the concentration of solutions.
The Eclipse professional-grade range of scales are available for general use in a variety of applications, with special models available, such as Low Volume Brix models to facilitate measurement of samples of less than 1μL that are used by botanists and zoologists for measuring nectar. All instruments are supplied with a soft carrying case and instructions for use.
These reliable and robust refractometers are sealed against the ingress of water with IP65 construction, so they can be used in the harshest of environments. The large body housing and rubber handgrip make the instrument incredible easy to handle. Special features make measurements easy and reliable:
  • Simple zero adjust with lock
  • Shaded view with bright display for maximum distinction
  • High precision, clear scale
  • Easy-clean prism
  • Unique sample ‘dribble feed’ feature
  • Low weight
  • Anti-roll supports
  • Push-on prism flap
TRUSTED_SUSTAINABILITY
Specifications
Type Eclipse Professional Grade Optical Hand Held Refractometer
Refractive Index 1.330 to 1.420
Increments 0.2
Scale Type Refractive Index
For Use With (Application) ‘On the spot’ determination of Refractive Index (RI) of liquids or the concentration of solutions
Includes Soft carrying case
IP Rating IP65