Learn More
Labcon™ Eclipse™ 300 μL Pipette Tips
These pipette tips fit most popular 300 μL pipettors.
Brand:  Labcon™ 1039-165-000-9
Additional Details : Weight : 0.34470kg
 View more versions of this product
Product Code. 17587796
  • 577.00 NOK / Pack of 200
Estimated Shipment: 24-11-2023
Description
  • Beveled point
  • If sterile, Validated Sterilization (ISO 11137) for use in Research and Medical Labs
  • Polypropylene USP Class VI Resin
  • Made in USA
Specifications
Specifications
300 μL
Sterile
Autoclavable
52.1 mm
ATP, RNase/DNase, Endotoxin-free
E-beam
Beveled
200 Tips/Pk., 10 Pk./Cs.
Yes
Eclipse™
Non-Filtered
Clear
2.05 in.
ISO 9001:2015, FDA CFR21 GMP, USP Class VI
Liquid Handling
Individually Wrapped
1 Tip/Bag, 200 Bags/Pk. 10 Pk./Cs.
Manufactured with 93% Renewable Energy
No
Polypropylene Resin