Learn More
Labcon™ Eclipse™ 200 μL Beveled Point Yellow Pipette Tips
GREENER_CHOICE
Product Code. 16683882
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Format:
Bagged
Bagged (1000)
Bagged (30,000)
Mini Refill
Rack, 96
Refill
Stack Rack
Unit Size:
Pack of 1000
Pack of 200
Pack of 30000
Pack of 480
Pack of 960
Product Code. Sterility Format unitSize
16683882 Sterile Refill Pack of 960
16663882 Sterile Bagged Pack of 200
16673882 Non-sterile Refill Pack of 960
16693882 Non-sterile Bagged (1000) Pack of 1000
16603892 Non-sterile Bagged (30,000) Pack of 30000
16661903 Non-sterile Rack, 96 Pack of 960
16671903 Sterile Rack, 96 Pack of 960
17577786 Non-sterile Mini Refill Pack of 480
17587786 Non-sterile Stack Rack Pack of 960
17597786 Non-sterile Stack Rack Pack of 960
17507796 Non-sterile Stack Rack Pack of 960
17517796 Sterile Stack Rack Pack of 960
17527796 Non-sterile Stack Rack Pack of 960
13 options
Product Code. 16683882
Brand: Labcon™ 10302650009
Only null left
Add to basket
Add to basket
These pipette tips fit most popular 200 μL pipettors.
  • Beveled point
  • ATP, RNase/DNase and Endotoxin-free (non-pyrogenic)
  • Polypropylene USP Class VI Resin
  • Made in USA
Specifications
Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Sterile
Color Yellow
Autoclavable Autoclavable
Length (English) 1.95 in.
Length (Metric) 49.7 mm
Certifications/Compliance ISO 9001:2015; FDA CFR21 GMP
Clean Claims ATP, RNase/DNase, and Endotoxin-free
For Use With (Application) Liquid Handling
Sterilization Method E-beam
Format Refill
Tip Style Beveled
Quantity 960 Tips/Refill, 10 Refills/Cs.
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated No
Product Line Eclipse
Material Polypropylene Resin