Learn More
Labcon Eclipse™ 10 μL Extra Long Graduated Pipette Tips with UltraFine™ Point
Product Code. p-7194059
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Format:
Bagged
Rack
Unit Size:
Pack of 1000
Pack of 960
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Format unitSize
16623902 Non-sterile Bagged Pack of 1000
16633902 Non-sterile Rack Pack of 960
16643902 Sterile Rack Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16623902 Supplier Labcon Supplier No. 10378000009
Only null left
Add to basket
Add to basket
Fits Popular 10 μL Pipettors
  • Pipette Tip
  • Eclipse™ Extra Long
  • Graduated at 2 μL and 10 μL
  • With UltraFine™ point
  • Fits Popular 10 μL Pipettors
  • ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
  • Polypropylene USP Class VI resin
  • Made in USA
Specifications
Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Length (English) 1.87 in.
Length (Metric) 47.6 mm
Certifications/Compliance ISO 9001:2015; FDA CFR21 GMP
Clean Claims ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
For Use With (Application) Liquid Handing
Format Bagged
Tip Style UltraFine
Packaging 1000 Tips/Bag, 10 Bags/Cs.
Quantity 1000 tips/bag
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line Eclipse
Material Polypropylene Resin