missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ECHDC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27732-1-AP-20UL
This item is not returnable.
View return policy
Description
ECHDC1 belongs to the enoyl-CoA hydratase/isomerase family. The exact function of ECHDC1 remains unknown.Specifications
| ECHDC1 | |
| Polyclonal | |
| Unconjugated | |
| ECHDC1 | |
| ECHDC1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 361465, 52665, 55862 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6AYG5, Q9D9V3, Q9NTX5 | |
| Echdc1 | |
| ECHDC1 Fusion Protein Ag26905 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction