Learn More
ECE2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15418-1-AP-150UL
This item is not returnable.
View return policy
Description
Converts big endothelin-1 to endothelin-1. Also involved in the processing of various neuroendocrine peptides, including neurotensin, angiotensin I, substance P, proenkephalin-derived peptides, and prodynorphin-derived peptides. May limit beta-amyloid peptide accumulation in brain. May also have methyltransferase activity.Specifications
| ECE2 | |
| Polyclonal | |
| Unconjugated | |
| ECE2 | |
| ECE2, endothelin converting enzyme 2, KIAA0604 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 107522, 408243, 9718 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O60344, Q80Z60 | |
| Ece2 | |
| ECE2 Fusion Protein Ag7706 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |