Learn More
Terumo™ Luer-Lock Tip Three-Part Syringe
Product Code. 11866071
Change view
Click to view available options
Volume (Metric):
20 mL
Quantity:
50 Pk.
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Quantity Unit Size
11866071 20 mL 50 Pk. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11866071 Supplier Terumo™ Supplier No. 8SS20L1
  • ONLINE EXCLUSIVE PRICE
    697.20 NOK
    109.70 NOK / Pack of 50
    Save 587.50 NOK
    84% Off
 Restricted Item
Estimated Shipment: 23-11-2026
Terumo™ Luer-Lock Tip Three-Part Syringes are easy to use with their smooth plunger action and have clear barrels to allow quick recognition of air bubbles for more accurate dosing. With easy to read markings, you can be confident you are delivering the intended dose every time.
Use these syringes for the aspiration of fluids or sampling blood or other body fluids. Terumo™ Luer-Lock Three-Part Syringes feature a unique double beveled design for greater patient comfort.
  • Silicone coated barrel for a smooth plunger action
  • Clear barrel to allow quick recognition of air bubbles
  • Easy to read markings
  • Latex free
  • Sterile and packed in individually wrapped tamper peel packs
  • Syringes are available with luer lock, luer slip, catheter and eccentric tips
  • Available in a wide range of sizes
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Certifications/Compliance Class IIa, sterile product following MDD 93/42/EEC, CE 0197
Material Polypropylene
Description Screen printed graduation
For Use With (Application) For the injection of fluids immediately after filling into the human body, for aspiration of fluids or for the sampling of blood or other body fluids
Latex Free Yes
Product Type Luer-Lock Tip Three-Part Syringe, Concentric
Disposable Single-use
Quantity 50 Pk.
Volume (Metric) 20 mL
Connection Type Luer Lock
Graduated Yes