Learn More
Trajan™ EC GC Septa
Product Code. p-8010100
Click to view available options
Diameter (Metric) Outer:
6 mm
7 mm
8 mm
9 mm
10 mm
11 mm
17 mm
Temperature (Metric) Max:
300°C
350°C
Unit Size:
Pack of 25
Pack of 50
Product Code. 10330421
Brand: Trajan™ 041842
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
EC GC septa is designed for long lifetime and low injection port adhesion
  • Combines long injection lifetime, low bleed and low injection port adhesion
  • Reliable seal against carrier gas pressure in the inlet
  • No bleed or contamination into the GC system
  • Maximum operating temperature of 350°C (300°C for 17mm size)
Specifications
Type Long Life EC Septum
Diameter (Metric) Outer 7 mm
Temperature (Metric) Max 350°C
Material High Temperature Silicon
Quantity 50/Pk.