Learn More
Invitrogen™ eBioscience™ Human Lysed Whole Blood Cell Flow Cytometry Workflow Kit
Product Code. 17797463
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17797463 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17797463 Supplier Invitrogen™ Supplier No. A53013
The eBioscience™ Human Lysed Whole Blood Cell Flow Cytometry Workflow Kit contains all of the normal buffers, blockers, and beads needed to complete a standard flow cytometry experiment for staining cell surface targets on human lysed whole blood.
  • Kit contains all of the normal buffers, blockers, and beads to complete a standard flow cytometry experiment for staining cell surface targets on human lysed whole blood
  • When combined with experiment-specific antibodies, the reagents within this kit have been optimized to work together to detect human blood cell surface reagents by flow cytometry
  • Effective compensation controls
  • FC receptor blocking
  • Non-antibody, non-protein based blocking solution for reducing background staining from fluorophores
  • Antibody binding buffer maximizes non-specific binding of antibodies
  • Fixation buffer for storage of stained samples
Kit Includes
  • UltraComp eBeads™ Plus Compensation Beads (01-3333-41)
  • eBioscience 10X RBC Lysis Buffer, multi-species (00-4300-54)
  • Fc Receptor Binding Inhibitor Polyclonal Antibody, eBioscience (14-9161-71)
  • CellBlox™ Blocking Buffer (B001T06F01)
  • eBioscience™ Flow Cytometry Staining Buffer (00-4222-57)
  • eBioscience™ IC Fixation Buffer (00-8222-49)
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
For Use With (Application) Flow Cytometry