Learn More
Ebastine, TRC
Product Code. 30008082
Change view
Click to view available options
Quantity:
10 mg
500 mg
1 g
Unit Size:
10mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008082 500 mg 500mg
17978504 1 g 1g
17978984 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30008082 Supplier Toronto Research Chemicals Supplier No. TRCE320000500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 90729-43-4
Molecular Formula C32 H39 N O2
Molecular Weight (g/mol) 469.66
Synonym Ebastine, 1-[4-(1,1-Dimethylethyl)phenyl]-4-[4-(diphenylmethoxy)-1-piperidinyl]-1-butanone, Bastel, Ebastel, Ebastin, LAS-W 090, RP 64305
IUPAC Name 4-(4-benzhydryloxypiperidin-1-yl)-1-(4-tert-butylphenyl)butan-1-one
SMILES CC(C)(C)c1ccc(cc1)C(=O)CCCN2CCC(CC2)OC(c3ccccc3)c4ccccc4
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C32H39NO2/c1-32(2,3)28-18-16-25(17-19-28)30(34)15-10-22-33-23-20-29(21-24-33)35-31(26-11-6-4-7-12-26)27-13-8-5-9-14-27/h4-9,11-14,16-19,29,31H,10,15,20-24H2,1-3H3
Formula Weight 469.2981
Percent Purity >95%
Chemical Name or Material Ebastine
Grade HPLC
Product Title
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.