Learn More
STEMCELL Technologies EasySep™ Red Blood Cell Lysis Buffer
Product Code. p-7217690
Encompass_Preferred
Click to view available options
Quantity:
10 mL
100 mL
Unit Size:
100mL
10mL
Product Code. 17108021
Brand: STEMCELL Technologies 20110
Red blood cell lysis buffer, 10X concentrate
  • EasySep™ Red Blood Cell Lysis Buffer is for use as directed with EasySep™ or RoboSep™ Whole Blood kits for the lysis of red blood cells prior to separation from whole blood
Specifications
Description Chimerism; HLA; Immunology
Quantity 10 mL
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.