Learn More
Ansell™ Easy Flex 47-200 Series Nitrile Medium Weight Gloves
Product Code. 11796733 Shop All Ansell Products
Change view
Click to view available options
Size:
6.5
7
8
9
10
Unit Size:
12 pairs
Product Code. Size unitSize
11796733 8 12 pairs
11716743 10 12 pairs
12610902 6.5 12 pairs
12620902 7 12 pairs
11706743 9 12 pairs
5 options
Product Code. 11796733
Brand: Ansell™ 47200080
Only null left
Add to basket
Add to basket
The coolness and comfort of cotton - but with far superior grip. Coating only on fingers and palm to protect the working area, but leaves the back of the hand free to breathe.
  • Much better resistance to snags, puncture and abrasion.
  • Nitrile coating repels oil and grease
  • Safely handle smooth objects like boxes or appliances with ease
  • Lasts five times longer than an ordinary cotton glove
  • Cut-and-sewn construction
  • Interlock cotton liner
  • 3/4 Dipped
  • Knitwrist cuff
TRUSTED_SUSTAINABILITY
Specifications
Back Material Nitrile
Certifications/Compliance CE Mark, ANSI, CATEGORY II EN 388 : 2111
Cuff Material Nitrile
Coating Material Nitrile
Color Green
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Cotton
Product Type Mediumweight Gloves
Size 8
Glove Cuff Style Knit
Liner Material Interlock Cotton
Lining Lined
Palm Type Coated-palm
Quantity 144 Pairs
Resistant To Snag, puncture, abrasion and cut resistance
Series 47-200