Learn More
Hoefer™ Easy Breeze™ Gel Dryer Accessories: Frame Components and Accessories
Product Code. p-4895496 Shop All Cytiva Products
Click to view available options
Type:
Quarter-turn Knob
Unit Size:
Pack of 10
Product Code. 11505604
Brand: Hoefer™ SE1213
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For use with the SE 1200 Easy Breeze Gel Drying System
  • Highly resistant to acids, bases, and all commonly used buffer systems
  • Easily removed while maintaining gel integrity
  • Lightweight and virtually unbreakable when used with reasonable care
TRUSTED_SUSTAINABILITY
Specifications
Color Black
For Use With (Equipment) SE 1200 Easy Breeze Gel Drying System
Type Quarter-turn Knob