Learn More
3M™ Earplugs Top Up Bag
Product Code. 17297640 Shop All 3M Products
Change view
Click to view available options
37 dB
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17297640 37 dB
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17297640 Supplier 3M™ Supplier No. 7100100263
  • ONLINE EXCLUSIVE PRICE
    6200.00 NOK
    3692.01 NOK / 2000 pairs
    Save 2507.99 NOK
    40% Off
Estimated Shipment: 07-12-2026
Earplugs made from soft, hypoallergenic foam. They can reduce noise levels by 37 dB and have a tapered design that fits comfortably in your ears.
  • Reduces noise levels by 37 dB
  • Model is compatible for use with the 3M™ E-A-RFit™ Validation system and available as a probed plug
  • Tapered design fits comfortably in your ears
  • Soft, hypoallergenic foam provides minimum pressure in the ear
  • 2000 pairs per box
  • Available with probed feature for use with 3M™ E-A-RFit™ Dual-Ear Validation System
TRUSTED_SUSTAINABILITY
Specifications
Class 1100 R
Quantity 2000 Pairs/Box
Noise Reduction Rating (NRR) 37 dB
Corded Uncorded