Learn More
Sperian™ Howard Leight™ Bilsom™ 303 Disposable Earplugs
Product Code. 15221149
Click to view available options
:
Color: Yellow and White
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Product Code. 15221149
Brand: Sperian™ 1006187
Only null left
Add to basket
Add to basket
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Wears more comfortably than other foam earplugs over long periods of time. Sperian™ Howard Leight™ Bilsom™ 303 Disposable Earplugs are ideal for medium- to high-noise environments.
  • Polyurethane foam expands gently in ear canal, resulting in less expansion pressure
  • Smooth, soil-resistant skin prevents dirt build-up
  • Tapered bullet-shaped design is easier to roll-down and insert, adapts more evenly to ear canal
TRUSTED_SUSTAINABILITY
Specifications
Color Yellow, White
Size Universal
Quantity 1 Ea.
Plug Shape Bullet
Product Type Earplug