missing translation for 'onlineSavingsMsg'
Learn More

EAI045, MedChemExpress

Product Code. p-200096424
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16474256

Brand: Medchem Express HY100213

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress EAI045 is an allosteric and the fourth-generation inhibitor of mutant EGFR with IC50s of 1.9, 0.019, 0.19 and 0.002 μM for EGFR, EGFRL858R, EGFRT790M and EGFRL858R/T790M at 10 μM ATP, respectively.

EAI045 is an allosteric and the fourth-generation inhibitor of mutant EGFR with IC50s of 1.9, 0.019, 0.19 and 0.002 μM for EGFR, EGFRL858R, EGFRT790M and EGFRL858R/T790M at 10 μM ATP, respectively.

Specifications

Chemical Name or Material EAI045
CAS 1942114-09-1
Color off White
Purity Grade Notes Research
Molecular Formula C19H14FN3O3S
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (260.82 mM; Need ultrasonic)
SMILES O=C(C(C1=C(O)C=CC(F)=C1)N2C(C(C=CC=C3)=C3C2)=O)NC4=NC=CS4
Molecular Weight (g/mol) 383.4
Formula Weight 383.4
Percent Purity 96.53%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.