missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
EAAT3 Polyclonal antibody specifically detects EAAT3 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | EAAT3 |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 450 SE |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EAAC1excitatory amino acid carrier 1, EAAT3excitatory amino acid transporter 3, Excitatory amino-acid carrier 1, Neuronal and epithelial glutamate transporter, Sodium-dependent glutamate/aspartate transporter 3, solute carrier family 1 (neuronal/epithelial high affinity glutamatetransporter, system Xag), member 1, Solute carrier family 1 member 1 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 430-524 of human EAAT3 (NP_004161.4).,, Sequence:, SIKPGVTQKVGEIARTGSTPEVSTVDAMLDLIRNMFPENLVQACFQQYKTKREEVKPPSDPEMNMTEESFTAVMTTAISKNKTKEYKIVG |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?