Learn More
EAAT2 Mouse anti-Human, Unconjugated, Clone: 1C8D10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67083-1-IG-150UL
Description
The EAAT2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EAAT2. This antibody recognizes human, mouse, rat antigen. The EAAT2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| EAAT2 | |
| Monoclonal | |
| 800 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunohistochemistry | |
| 1C8D10 | |
| Unconjugated | |
| 150 μL | |
| 6506 | |
| IgG1 |