missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E2A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | E2A |
---|---|
Concentration | 0.6 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16890855
|
Proteintech
21242-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16880855
|
Proteintech
21242-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
TCF3 (E2A) is a transcription factor that plays major roles in determining tissue-specific cell fate during embryogenesis, like muscle or early B-cell differentiation. Heterodimers between E2A and tissue-specific basic helix-loop-helix (bHLH) Dimers bind DNA on E-box motifs: 5′- CANNTG-3′. Binds to the kappa-E2 site in the kappa immunoglobulin gene enhancer. Deletions in E2A have been observed in a subset of pre-B-cell acute lymphoblastic leukemia (B-ALL) cases. Two alternatively spliced human isoforms have been described.Specifications
E2A | |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P15806, P15923, P21677 | |
171046, 21423, 6929 | |
E2A Fusion Protein Ag15734 | |
Primary | |
-20°C | |
TCF3 |
0.6 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
bHLHb21, E2A, E47, ITF1, Kappa E2 binding factor, TCF 3, TCF3, Transcription factor 3, Transcription factor E2 alpha, Transcription factor ITF 1 | |
Tcf3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |