Learn More
Invitrogen™ E-Gel™ EX Mix Pack
Description
- These pre-cast agarose gels are designed for ultrasensitive and convenient DNA sample electrophoresis in as little as eight minutes
- Detect 1 ng of sample DNA
- Complete DNA sample separation in eight minutes
- Openable cassette design allows easy in-gel access for convenient DNA sample excision or transfer to membranes
- Precast E-Gel agarose gels are ready for use and do not require gel preparation or additional buffers
- E-Gel agarose gels do not require buffers to run
Mix Pack includes:
- E-Gel EX Agarose Gels, 1%
- E-Gel EX Agarose Gels, 2%
- E-Gel EX Double Comb Agarose Gels, 1%
- E-Gel EX Double Comb Agarose Gels, 2%
- E-Gel SizeSelect II Agarose Gels, 2%
Running and imaging devices
- E-Gels EX agarose gels require the E-Gel Power Snap or E-Gel Power Snap Plus electrophoresis systems for gel running and viewing
- The systems consist of an electrophoresis device and a camera for fast and convenient E-Gel agarose gel separation and analysis
Specifications
Specifications
| Format | Precast Gel |
| Gel Type | E-Gel |
| Separation of | DNA |
| Separation Type | Electrophoresis |
| Wells | 11-well |
| Quantity | 10 Gels |
| Sterility | Non-sterile |
For Research Use Only.