Learn More
E,E-Farnesylacetone, TRC
Product Code. 30403432
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30403432 5 g
30401858 1 g
30402649 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30403432 Supplier Toronto Research Chemicals Supplier No. TRCF1024455G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1117-52-8
Molecular Formula C18 H30 O
Molecular Weight (g/mol) 262.43
Synonym Teprenone Impurity 16
IUPAC Name (5E,9E)-6,10,14-trimethylpentadeca-5,9,13-trien-2-one
SMILES CC(=CCC\C(=C\CC\C(=C\CCC(=O)C)\C)\C)C
Specifications
Quantity 5 g
InChI Formula InChI=1S/C18H30O/c1-15(2)9-6-10-16(3)11-7-12-17(4)13-8-14-18(5)19/h9,11,13H,6-8,10,12,14H2,1-5H3/b16-11+,17-13+
Formula Weight 262.2297
Chemical Name or Material E,E-Farnesylacetone