missing translation for 'onlineSavingsMsg'
Learn More

Scharlab E. Coli Broth

Product Code. 11401787
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11401787 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11401787 Supplier Scharlab Supplier No. 02060500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

E. Coli Broth is part of a range of selective broths for Enterobacteriaceae. Its efficiency or selectivity is based on bile salts' inhibitory effect on other microorganisms.

  • APHA/COMPF/ISO/SMWW/WEF
  • Selective medium for the detection and enumeration (MPN) of enterobacteria, in water and foodstuffs
  • According to ISO 7251 and 9308-2 standards.

Specifications

Certifications/Compliance ISO 7251, 9308-2
Form Liquid
Media Type E. Coli Broth
Product Type Microbiology Media
Target Organism Class Gram-negative Bacteria
Formulation Peptone 20.0 g/L, Bile Salts No. 3 1.5 g/L, Lactose 5.0 g/L, Dipotassium Phosphate 4.0 g/L, Potassium Dihydrogen Phosphate 1.5 g/L, Sodium Chloride 5.0 g/L
For Use With (Application) Detection and Enumeration of Enterobacteria
Incubation Conditions 44.5°C ± 0.5°C
Incubation Time 21 ±3 hr.
pH 6.9 ±0.2
Quantity 500 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.