Learn More
3M™ E-A-R™ UltraTech Earplugs
Product Code. 17217630
Change view
Click to view available options
:
Color: Black, White
Product Code.
17217630 Color: Black, White
1 options
Product Code. 17217630
Brand: 3M™ 7000103799
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
3M™ E-A-R™ UltraTech Earplugs greatly improve the ability to perceive speech, warning signals and machinery noise whilst reducing harmful noise levels effectively.
  • Improve wearer's ability to perceive speech, warning signals and machinery noise whilst reducing harmful noise levels effectively
  • Earplug are ideal for musicians who require a flat attenuation protection
  • Conical shape plugs help provide effective seal for reassured protection
  • Effective protection against moderate levels of harmful noise SNR 21 dB
TRUSTED_SUSTAINABILITY
Specifications
Color Black, White
Pairs per Package 1 Pair/Box
Noise Reduction Rating (NRR) 21 dB
Corded Uncorded
Dielectric No