Learn More
3M™ E-A-R™ Push-Ins Earplugs
Product Code. p-7160291
Click to view available options
Corded:
Corded
Uncorded
Unit Size:
100 pairs
50 pairs
Product Code. 15739777
Brand: 3M™ 7000052713
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
3M™ E-A-R™ Push-Ins Earplugs feature a unique E-A-Rform™ foam tip that is shaped and sized to mold comfortably to fit most ear canals.
  • With Push-Ins earplugs a gentle push is all it takes for easy, consistent fitting
  • No need to handle the plug tip making them more hygienically acceptable
  • Foam tip is shaped and sized for easier fitting
  • Blue flexible stem makes fitting and removal easy and helps keeps the tips clean when the wearer's hands are dirty
TRUSTED_SUSTAINABILITY
Specifications
Color Yellow, Blue
Certifications/Compliance CE Approved
Packaging Type Polybag
Pairs per Package 400 Pairs/Box
Noise Reduction Rating (NRR) 38 dB
Corded Corded
Product Type Push-In Earplugs
Dielectric No
Material Polyurethane