Learn More
3M™ E-A-R™ Classic™ Soft™ Earplugs PR-01-005
Description
- Earplugs starts out firm to allow for easy fitting then softens from body heat for a comfortable fit
- Proprietary thermal-reactive foam
- Overtime the foam continues to soften for added comfort
- Provides an excellent seal due to the shaping of the earplug to the ear canal
- Textured surface helps keep earplugs in place
Specifications
Specifications
| Color | Yellow |
| Certifications/Compliance | CE Approved |
| Packaging Type | Polybag |
| Pairs per Package | 200 Pairs/Box |
| Noise Reduction Rating (NRR) | 36 dB |
| Corded | Corded |
| Product Type | Earplugs |
| Dielectric | No |
| Material | Slow-recovery PVC Foam |