Learn More
(E)-3-(5-Nitrofuran-2-yl)acrylaldehyde, TRC
Product Code. 30481774
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481774 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30481774 Supplier Toronto Research Chemicals Supplier No. TRCN495950250MG
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (E)-3-(5-Nitrofuran-2-yl)acrylaldehyde
CAS 52661-56-0
Molecular Formula C7H5NO4
Quantity 250 mg
Synonym (2E)-3-(5-Nitro-2-furanyl)-2-propenal, (E)-3-(5-Nitro-2-furyl)acrolein, trans-3-(5-Nitro-2-furyl)acrolein
InChI Formula InChI=1S/C7H5NO4/c9-5-1-2-6-3-4-7(12-6)8(10)11/h1-5H/b2-1+
SMILES C(=C\C1=CC=C([N+](=O)[O-])O1)/C=O
IUPAC Name (E)-3-(5-nitrofuran-2-yl)prop-2-enal
Molecular Weight (g/mol) 167.12
Formula Weight 167.0219