Learn More
Dysprosium Oxide, Sigma-Aldrich™
Specifications
Specifications
| Melting Point | 2340°C |
| CAS | 1308-87-8 |
| Quantity | 10 g |
| Density | 7.81 g/cm3 at 20°C |
| MDL Number | MFCD00010986 |
| Synonym | Dysprosium oxide, Dysprosium oxide |
| Molecular Weight (g/mol) | 373 |
| Grade | Research |
| Chemical Name or Material | Dysprosium Oxide |