missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYNLT3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | DYNLT3 |
|---|---|
| Concentration | 0.3 mg/mL |
| Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16858933
|
Proteintech
11687-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16848933
|
Proteintech
11687-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| DYNLT3 | |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| P51808, P56387 | |
| 363448, 67117, 6990 | |
| DYNLT3 Fusion Protein Ag2299 | |
| Primary | |
| -20°C | |
| DYNLT3 |
| 0.3 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| DYNLT3, Protein 91/23, RP3, TCTE1L, TCTE1XL, TCTEX1L | |
| Dynlt3 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title