missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYNC2LI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | DYNC2LI1 |
|---|---|
| Concentration | 0.25 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16855354
|
Proteintech
15949-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16845354
|
Proteintech
15949-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DYNC2LI1 (dynein, cytoplasmic 2, light intermediate chain 1), also known as LIC3, D2LIC or CGI-60, is a 351 amino acid cytoplasmic protein belonging to the dynein light intermediate chain family. DYNC2LI1 may function as a motor for intraflagellar retrograde transport and in cilia biogenesis. The cytoplasmic dynein complex 2 may be composed of a DYNC2H1 homodimer and a number of DYNC2LI1 light intermediate chains. DYNC2LI1 exists as five alternatively spliced isoforms.Specifications
| DYNC2LI1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat, Human | |
| Q6AY43, Q8K0T2, Q8TCX1 | |
| 213575, 298767, 51626 | |
| DYNC2LI1 Fusion Protein Ag8584 | |
| Primary | |
| -20°C | |
| DYNC2LI1 |
| 0.25 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CGI 60, D2LIC, DYNC2LI1, LIC3 | |
| DYNC2LI1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title