missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DYNC2LI1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15912705
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15912705 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15912705 Supplier Invitrogen™ Supplier No. PA576175

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

DYNC2LI1 (dynein, cytoplasmic 2, light intermediate chain 1), also known as LIC3, D2LIC or CGI-60, is a 351 amino acid cytoplasmic protein belonging to the dynein light intermediate chain family. DYNC2LI1 may function as a motor for intraflagellar retrograde transport and in cilia biogenesis. The cytoplasmic dynein complex 2 may be composed of a DYNC2H1 homodimer and a number of DYNC2LI1 light intermediate chains. DYNC2LI1 exists as five alternatively spliced isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DYNC2LI1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Dync2li1
Gene Accession No. Q6AY43, Q8K0T2, Q8TCX1
Gene Alias 4933404O11Rik; CGI-60; Cytoplasmic dynein 2 light intermediate chain 1; D2LIC; Dync2li1; Dynein 2 light intermediate chain; dynein cytoplasmic 2 light intermediate chain 1; dynein, cytoplasmic 2, light intermediate chain 1; LIC3; mD2LIC; RGD1310286
Gene Symbols Dync2li1
Host Species Rabbit
Immunogen Recombinant fusion protein of human DYNC2LI1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 213575, 298767, 51626
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.