Learn More
ABEBA™ Dynamic Flow Anthracite Occupational Slip-On O1
Product Code. 17101635
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 17101635
Brand: ABEBA™ 7375/48
Only null left
Add to basket
Add to basket
Occupational slip-on shoes, breathable anthracite knitted fabric, replaceable active MediFit insole, slip resistant TPU outsole
  • Breathable, lightweight, comfortable
  • Comply to EN ISO 20347: 2012, O1 SRC
  • Slip resistant TPU outsole
  • Good cushioning
  • Energy absorption in heel area
  • Anthracite knitted fabric breathable
  • Double elastic band fitting system
  • Removable active Medifit insole
Specifications
Color Anthracite
Material Knitted Fabric
Size 48
Certifications/Compliance (Europe) Comply to EN ISO 20347: 2012 O1 SRC
Gender Unisex
Ankle Style Slip-on
Waterproof Water Repellent
Product Type O1 SRC ESD