Learn More
ABEBA™ Dynamic Blue Occupational Clogs Ob ESD
Product Code. 17121325
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 17121325
Brand: ABEBA™ 37326/38
Only null left
Add to basket
Add to basket
Blue dynamic occupational clog with non-foldable heel strap, replaceable active comfort insole, slip resistant TPU outsole
  • Breathable, lightweight, comfortable occupational clogs
  • Comply to EN ISO 20347: 2012, OB SRC
  • ESD dissipative according to DIN EN 61340
  • Slip resistant TPU outsole
  • Good cushioning
  • Energy absorption in heel area
  • Water repellent, blue knitted textile
  • Non foldable heel strap, adjustable
  • Removable active comfort insole
Specifications
Color Blue
Material Knitted Fabric
Size 38
Certifications/Compliance (Europe) ESD dissipative to EN 61340 Climate class 1, Comply to EN ISO 20347: 2012 OB SRC
Gender Unisex
Ankle Style Clogs
Waterproof Water Repellent
Product Type OB SRC ESD