Learn More
Applied Biosystems™ Dynabeads™ Rotary Mixer Mixing Platform
Product Code. 10338784 Shop All Applied Biosystems Products
Click to view available options
For Use With (Equipment):
Dynabeads™ Sample Mixers
Unit Size:
Each
Product Code. 10338784
Brand: Applied Biosystems™ 94703
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Dynabeads™ Rotary Mixer Mixing Platform
When attached to the Dynabeads™ Rotary Mixer (Cat. No. 947-01), the Mixing Platform can hold Dynabeads in 3 MPC™-S racks or one 96-well microcentrifuge tube holder provided with the Mixing Platform. Variable speed (15-80 rpm). Can be operated in both cold and warm conditions (4-37°C).
TRUSTED_SUSTAINABILITY
Specifications
Speed Range 15 to 80 rpm
For Use With (Equipment) Dynabeads™ Sample Mixers
Product Line Dynabeads, MPC
For Research Use Only. Not for use in diagnostic procedures.