Learn More
Applied Biosystems™ Dynabeads™ Rotary Mixer Mixing Platform
Shop All Applied Biosystems Products
Click to view available options
For Use With (Equipment):
Dynabeads™ Sample Mixers
Unit Size:
Each
Description
When attached to the Dynabeads™ Rotary Mixer (Cat. No. 947-01), the Mixing Platform can hold Dynabeads in 3 MPC™-S racks or one 96-well microcentrifuge tube holder provided with the Mixing Platform. Variable speed (15-80 rpm). Can be operated in both cold and warm conditions (4-37°C).
Specifications
Specifications
| Speed Range | 15 to 80 rpm |
| For Use With (Equipment) | Dynabeads™ Sample Mixers |
| Product Line | Dynabeads, MPC |