Learn More
Applied Biosystems™ Dynabeads™ MPC™-S (Magnetic Particle Concentrator)
Shop All Applied Biosystems ProductsDescription
The MPC™-S was designed specifically for microbiology applications; it has been validated with select Dynabeads™ for enrichment of E. coli O157:H7, Salmonella, non-O157 STEC, Cryptosporidium, and Giardia. The MPC™-S can be used with both food enrichment samples types as well as water samples. It holds up to six microcentrifuge tubes and has an optimal working volume range of 20 μl–2 ml.
Note that this magnet is for microbiology applications only, not for eukaryotic cell separations. For use in food and environmental testing only. Not for any animal or human therapeutic or diagnostic use.
Specifications
Specifications
| Content And Storage | Store at room temperature. |
| Format | 6 Microtubes |
| For Use With (Equipment) | Dynabeads™ MPC™, Dynabeads™ Sample Mixers |
| High-throughput Compatibility | Not High-throughput Compatible (Manual) |
| Product Line | Dynabeads™ |
| Quantity | 1 unit |
| Sample Type | Bacterial Culture |
| Shipping Condition | Room Temperature |