missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ CD15

Product Code. 10252093
Change view
Click to view available options
Quantity:
2 mL
5 mL
Unit Size:
2mL
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10252093 5 mL Each
30275404 2 mL 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10252093 Supplier Invitrogen™ Supplier No. 11137D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dynabeads CD15 are superparamagnetic beads covalently coupled with an anti-human CD15 antibody that efficiently depletes human CD15+ myeloid cells directly from whole blood, buffy coat, or PBMC samples. Dynabeads CD15 can also be used to positively isolate CD15+ cells for molecular applications.

Dynabeads CD15 are superparamagnetic beads covalently coupled with an anti-human CD15 antibody that efficiently depletes human CD15+ myeloid cells (predominantly neutrophil and eosinophil granulocytes) directly from whole blood, buffy coat, or peripheral blood mononuclear cell (PBMC) samples. Dynabeads CD15 can also be used to positively isolate CD15+ cells for molecular applications. Advantages of Dynabeads CD15:

  • Efficient depletion of granulocytes (neutrophils and eosinophils)
  • Isolation of granulocytes directly from whole blood for molecular applications
  • No columns required

About Dynabeads CD15

Dynabeads CD15 are uniform, superparamagnetic polystyrene beads (4.5 μm diameter) coated with a monoclonal mouse IgM antibody recognizing the human CD15 membrane antigen. The CD15 antigen is predominantly expressed on human neutrophil and eosinophil granulocytes, and to a varying degree on monocytes.

Magnetic bead-based separation offers easy handling

Dynabeads CD15 is added to the sample under continuous mixing to optimize the binding of the Dynabeads to the target cells. By placing the sample on a magnet, the bead-bound target cells are separated from the rest of the sample in just 1 to 2 minutes. For depletion, remove the supernatant to a new tube for further studies and discard the bead-bound cells. For positive isolation for molecular studies, remove the supernatant and wash the bead-bound cells 2 to 3 times in buffer to get optimal purity. The cells can be lysed while still attached to the beads and the supernatant transferred to a new tube for downstream molecular analysis. Starting samples can be whole blood, PBMC, or buffy coat.

Automation-ready for use with KingFisher purification systems

The product is ideal for high-throughput workflows using KingFisher systems. A user guide and scripts for KingFisher Flex and Apex workflows are available for download on this page.

Learn more about Dynabeads products

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Granulocytes
Content And Storage This Product Contains: 5 mL Dynabeads™ CD15 coated with anti-CD15 monoclonal antibody.

Storage Conditions: 2°C to 8°C. Do not freeze.
Material Polystyrene
Product Type Antibody Coated Bead
Includes 5 mL Dynabeads CD15 coated with anti-CD15 monoclonal antibody.
Isolation Technology Depletion, positive isolation for molecular applications
No. of Cells Processes ∼2 x 109 cells total
Output Viability >95%
Product Line DYNAL, Dynabeads
Purity or Quality Grade Research Grade
Quantity 5 mL
Sample Type PBMC, Blood
Shipping Condition Room Temperature
Starting Material Cell No. 1 x 107 PBMCs per isolation
Target Species Human
Diameter (Metric) 4.5 μm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.