Learn More
Invitrogen™ DYDC2 Monoclonal Antibody (8G4)
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA522754
Product Code. 15569162
  • 4170.00 NOK / 100µg
Description
Peptide Sequence: METNYLKRCF GNCLAQALAE VAKVRPSDPI EYLAHWLYHY RKTAKAKEEN REKKIHLQEE YDSSLKEMEM TEMLKQEEYQ IQQNCEKCHK ELTSETVSTK KTIFMQEDTN PLEKEALKQE FLPGTSSLIP GMPQQVPPSE SAGQIDQNFK MPQEINYKEA FQHEVAHEMP PGSKSPF.
DYDC2 encodes a member of a family of proteins that contains a DPY30 domain. This gene locus overlaps with a closely related gene on the opposite strand. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DYDC2
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q96IM9
DYDC2
DYDC2 (AAH07374, 1 a.a. ~ 177 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2a κ
ELISA, Western Blot
8G4
Unconjugated
DYDC2
bA36D19.6; DPY30 domain containing 2; DPY30 domain-containing protein 2; DYDC2; MGC16186; SPS1; SRML1
Mouse
Affinity chromatography
RUO
84332
-20°C, Avoid Freeze/Thaw Cycles