missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYDC2 Antibody (8G4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00084332-M02
This item is not returnable.
View return policy
Description
DYDC2 Monoclonal antibody specifically detects DYDC2 in Human samples. It is validated for Western Blot, ELISA
Specifications
| DYDC2 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| bA36D19.6, DPY30 domain containing 2, DPY30 domain-containing protein 2, MGC16186 | |
| MGC16186 (AAH07374, 1 a.a. ∽ 177 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. METNYLKRCFGNCLAQALAEVAKVRPSDPIEYLAHWLYHYRKTAKAKEENREKKIHLQEEYDSSLKEMEMTEMLKQEEYQIQQNCEKCHKELTSETVSTKKTIFMQEDTNPLEKEALKQEFLPGTSSLIPGMPQQVPPSESAGQIDQNFKMPQEINYKEAFQHEVAHEMPPGSKSPF | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 8G4 | |
| Western Blot 1:500, ELISA | |
| AAH07374 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 84332 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction