missing translation for 'onlineSavingsMsg'
Learn More

Dyclonine hydrochloride, MP Biomedicals™

Product Code. 11471621 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
This item is not returnable. View return policy

Product Code. 11471621

Brand: MP Biomedicals 0219019425

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 536-43-6
Molecular Formula C18H28ClNO2
Molecular Weight (g/mol) 325.88
MDL Number MFCD00035386
InChI Key KNZADIMHVBBPOA-UHFFFAOYSA-N
Synonym dyclonine hydrochloride, dyclonine hcl, dyclone, dyclothane, tanaclone, 4'-butoxy-3-piperidinopropiophenone hydrochloride, dyclocaine hydrochloride, unii-zec193879q, 1-propanone, 1-4-butoxyphenyl-3-1-piperidinyl-, hydrochloride
PubChem CID 68304
ChEBI CHEBI:4725
IUPAC Name hydrogen 1-(4-butoxyphenyl)-3-(piperidin-1-yl)propan-1-one chloride
SMILES [H+].[Cl-].CCCCOC1=CC=C(C=C1)C(=O)CCN1CCCCC1

Specifications

Color White
Quantity 25 g
Formula Weight 325.9
Physical Form Powder
Chemical Name or Material Dyclonine hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.