Learn More
3M™ Dustmaster™ Particulate Filters
Particulate filter for use with the Dustmaster Powered Air Respirator helps protects against dust, mist and fume.
Brand: 3M™ 7000037674
655.00 NOK valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 0.12600kg
Description
For flexible P2 R protection against solid and liquid particles, use 3M™ Particulate Filters 5925. They add protection against particulates when used with 3M™ Gas and Vapour Filters, 6000 Series (using the 3M™ Filter Retainer). Our lightweight particulate filters help you work safely in hazardous environments. They’re designed to optimise your field of vision, and they can be used with any 3M™ Gas and Vapour filters, 6000 Series and 3M™ Reusable Masks that feature a bayonet-style connection.Specifications
EN143 : 2000, CE Approved | |
P2R | |
Solid and liquid particles | |
Filter |
Filter retainer | |
Particulate Filter | |
6000 and 7000 Series face masks |