Learn More
Pinnacle Brush™ Dusting Brushes
Product Code. p-8000733
Change view
Click to view available options
Length (Metric):
200 mm
240 mm
Unit Size:
Each
Product Code. Length (Metric) unitSize
12871924 240 mm Each
12861944 200 mm Each
2 options
Product Code. 12871924
Brand: Pinnacle Brush™ H1756
Metal-mounted brush. Pinnacle Brush™ Dusting Brushes are perfect for delicate tasks.
  • Metal mount
  • Wood handle
  • Fan shaped end
Specifications
Product Type Dusting Brush
Handle Material Wood
For Use With (Application) Dusting
Length (Metric) 240 mm
Quantity 1 Ea.