Learn More
Pinnacle Brush™ Dusting Brushes
Product Code. 12871924
Change view
Click to view available options
Length (Metric):
200 mm
240 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
12871924 240 mm Each
12861944 200 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12871924 Supplier Pinnacle Brush™ Supplier No. H1756
  • ONLINE EXCLUSIVE PRICE
    107.71 NOK
    71.00 NOK / Pack of 1
    Save 36.71 NOK
    34% Off
Estimated Shipment: 12-01-2027
Metal-mounted brush. Pinnacle Brush™ Dusting Brushes are perfect for delicate tasks.
  • Metal mount
  • Wood handle
  • Fan shaped end
Specifications
Product Type Dusting Brush
Handle Material Wood
For Use With (Application) Dusting
Length (Metric) 240 mm
Quantity 1 Ea.