Specifications
| Material | Durasil-25 |
| Particle Size | 5 to 17 μm |
| Pore Size | 60 Å |
| Quantity | 100 Pk. |
| Type | TLC Plate |
| Fluorescent Indicator | UV254 |
| Pore Volume | 0.75 mL/g |
| Specific Surface | ∼500 m2/g |
| Thickness (Metric) | 0.25 mm |
| Dimensions (L x W) | 5 x 20 cm |