Learn More
DWK Life Sciences DURAN™ Two Way Stopcock, double oblique bore, complete with glass key
Shop All DWK Life Sciences Products
Click to view available options
Plug Bore:
2.5 mm
4 mm
Unit Size:
Pack of 10
Description
DURAN™ patent valves complete with DURAN™ patent valve plugs are 2-way valves, which are frequently used for the construction of steering lines (vacuum inert gas lines, nitrogen distribution rakes). The tap plug is secured in the tap sleeve by a 3-part tap safety catch. We recommend the use of PRODURAN™ valves for particularly high leakage rate requirements.
- 2-way tap with threaded tap plug
- The tap plug is secured in the tap sleeve by a 3-part tap safety catch.
- Glass articles for further processing
Specifications
Specifications
| Plug Bore | 2.5 mm |
| Diameter (Metric) | 8 mm |
| Material | Borosilicate Glass (3.3), Polypropylene |
| Autoclavable | Autoclavable |
| Closure Size | 14.5 NS |
| Width (Metric) | 1.5 mm |
| No. per Pack | 10/Pk. |
| For Use With (Application) | Suitable for installation in glass apparatus for vacuum applications, e.g. for the construction of steering lines (vacuum inert gas lines, nitrogen distribution rakes) |