Learn More
Lenz Laborglasinstrumente™ Duran Tubing Reaction Vessels with Thermostatic Jacket
Description
DURAN™ tubing, cylindrical shape with graduation and laboratory flange LF. Thermostatic jacket with either GL 14, GL 18, KF 15 or KF 25 connectors.
Specifications
Specifications
| Capacity (Metric) | 4 L |
| Material | Borosilicate Glass, Duran Glass |
| Height (Metric) | 375 mm |
| Includes | Thermostatic jacket |
| Product Type | Reaction vessel |
| Shape | Cylindrical |
| Bottom Shape | Flat |