Learn More
Lenz Laborglasinstrumente™ Duran Tubing Reaction Vessels with Thermostatic Jacket
Product Code. 10616660
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
2000 mL
3 L
4 L
Height (Metric):
180 mm
190 mm
245 mm
275 mm
325 mm
375 mm
380 mm
Unit Size:
Each
Product Code. 10616660
Brand: Lenz Laborglasinstrumente™ 06010158
Only null left
Add to basket
Add to basket
Borosilicate glass reaction vessel with thermostatic jacket
DURAN™ tubing, cylindrical shape with graduation and laboratory flange LF. Thermostatic jacket with either GL 14, GL 18, KF 15 or KF 25 connectors.
Specifications
Capacity (Metric) 500 mL
Material Borosilicate Glass, Duran Glass
Height (Metric) 275 mm
Includes Thermostatic jacket
Product Type Reaction vessel
Shape Cylindrical
Bottom Shape Flat