Learn More
Lenz Laborglasinstrumente™ Duran Tubing Reaction Vessels with Thermostatic Jacket
Product Code. 10433360
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
2000 mL
3 L
4 L
Height (Metric):
180 mm
190 mm
245 mm
275 mm
325 mm
375 mm
380 mm
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Height (Metric) Unit Size
10433360 250 mL 190 mm Each
10391870 500 mL 180 mm Each
10616660 500 mL 275 mm Each
11532244 4 L 375 mm Each
11592234 2000 mL 375 mm Each
11512244 4 L 380 mm Each
11502244 3 L 325 mm Each
10349522 1000 mL 245 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10433360 Supplier Lenz Laborglasinstrumente™ Supplier No. 06010149
  • ONLINE EXCLUSIVE PRICE
    6025.00 NOK
    4739.25 NOK / Pack of 1
    Save 1285.75 NOK
    21% Off
Estimated Shipment: 25-01-2027
Borosilicate glass reaction vessel with thermostatic jacket
DURAN™ tubing, cylindrical shape with graduation and laboratory flange LF. Thermostatic jacket with either GL 14, GL 18, KF 15 or KF 25 connectors.
Specifications
Capacity (Metric) 250 mL
Material Borosilicate Glass, Duran Glass
Height (Metric) 190 mm
Includes Thermostatic jacket
Product Type Reaction vessel
Shape Cylindrical
Bottom Shape Flat