Learn More
Lenz Laborglasinstrumente™ Duran Tubing Reaction Vessels with Thermostatic Jacket
Borosilicate glass reaction vessel with thermostatic jacket
Brand: Lenz Laborglasinstrumente™ 06010258
Additional Details : Weight : 1.00000kg
Description
DURAN™ tubing, cylindrical shape with graduation and laboratory flange LF. Thermostatic jacket with either GL 14, GL 18, KF 15 or KF 25 connectors.Specifications
500 mL | |
180 mm | |
Reaction vessel | |
Flat |
Borosilicate Glass, Duran Glass | |
Thermostatic jacket | |
Cylindrical |