Learn More
Lenz Laborglasinstrumente™ Duran Tubing Reaction Vessels
Product Code. 11552164
Change view
Click to view available options
Height (Metric):
150 mm
200 mm
220 mm
300 mm
410 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
11552164 200 mm Each
11542174 410 mm Each
11542164 150 mm Each
11512184 220 mm Each
11502174 300 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11552164 Supplier Lenz Laborglasinstrumente™ Supplier No. 06000270
  • ONLINE EXCLUSIVE PRICE
    3200.00 NOK
    2664.11 NOK / Pack of 1
    Save 535.89 NOK
    16% Off
Estimated Shipment: 25-01-2027
Specifications
Capacity (Metric) 1000 mL
Material Borosilicate Glass, Duran Glass
Height (Metric) 200 mm
Product Type Reaction vessel
Shape Cylindrical
Bottom Shape Flat